RIE156322575 Rieke Metals, Inc.

4-(4-Cyanophenyl)-4-oxobutyronitrile

  • MDL number MFCD01319959
  • Cas Number : 898767-45-8
  • Molecular Weight : 184.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6022B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6022B
Availability details for
Quantity
  • Availability
  • Add to Cart