RIE156322576 Rieke Metals, Inc.

5-(4-Cyanophenyl)-5-oxovaleronitrile

  • MDL number MFCD01319960
  • Cas Number : 898767-48-1
  • Molecular Weight : 198.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6022C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6022C
Availability details for
Quantity
  • Availability
  • Add to Cart