RIE156322577 Rieke Metals, Inc.

6-(4-Cyanophenyl)-6-oxohexanenitrile

  • MDL number MFCD01319961
  • Cas Number : 898767-51-6
  • Molecular Weight : 212.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6022D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6022D
Availability details for
Quantity
  • Availability
  • Add to Cart