RIE156322578 Rieke Metals, Inc.

7-(4-cyanophenyl)-7-oxoheptanenitrile

  • MDL number MFCD02260560
  • Cas Number : 898767-54-9
  • Molecular Weight : 226.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6022E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6022E
Availability details for
Quantity
  • Availability
  • Add to Cart