RIE156322580 Rieke Metals, Inc.

4-(3-Cyanophenyl)-4-oxobutyronitrile

  • MDL number MFCD01319962
  • Cas Number : 898767-58-3
  • Molecular Weight : 184.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6023B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6023B
Availability details for
Quantity
  • Availability
  • Add to Cart