RIE156322581 Rieke Metals, Inc.

5-(3-cyanophenyl)-5-oxovaleronitrile

  • MDL number MFCD01319963
  • Cas Number : 898767-60-7
  • Molecular Weight : 198.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-cyanobutanoyl)benzonitrile
Inchi Code 1S/C12H10N2O/c13-7-2-1-6-12(15)11-5-3-4-10(8-11)9-14/h3-5,8H,1-2,6H2
Inchi Key YNEOODHLLFNBGN-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6023c

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6023c
Availability details for
Quantity
  • Availability
  • Add to Cart