RIE156322584 Rieke Metals, Inc.

8-(3-cyanophenyl)-8-oxooctanenitrile

  • MDL number MFCD02260563
  • Cas Number : 898767-66-3
  • Molecular Weight : 240.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(7-cyanoheptanoyl)benzonitrile
Inchi Code 1S/C15H16N2O/c16-10-5-3-1-2-4-9-15(18)14-8-6-7-13(11-14)12-17/h6-8,11H,1-5,9H2
Inchi Key JWIGARDAYDEYFO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6023f

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6023f
Availability details for
Quantity
  • Availability
  • Add to Cart