RIE156322585 Rieke Metals, Inc.

5-(3-chlorophenyl)-5-oxovaleronitrile

  • MDL number MFCD02260567
  • Cas Number : 898767-68-5
  • Molecular Weight : 207.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6026C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6026C
Availability details for
Quantity
  • Availability
  • Add to Cart