RIE156322586 Rieke Metals, Inc.

6-(3-chlorophenyl)-6-oxohexanenitrile

  • MDL number MFCD02260568
  • Cas Number : 898767-70-9
  • Molecular Weight : 221.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6026D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6026D
Availability details for
Quantity
  • Availability
  • Add to Cart