RIE156322589 Rieke Metals, Inc.

4-(2-Chlorophenyl)-4-oxobutyronitrile

  • MDL number MFCD02260571
  • Cas Number : 135595-17-4
  • Molecular Weight : 193.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6027B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6027B
Availability details for
Quantity
  • Availability
  • Add to Cart