RIE156322592 Rieke Metals, Inc.

7-(2-chlorophenyl)-7-oxoheptanenitrile

  • MDL number MFCD02260574
  • Cas Number : 898767-80-1
  • Molecular Weight : 235.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6027E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6027E
Availability details for
Quantity
  • Availability
  • Add to Cart