RIE156322594 Rieke Metals, Inc.

5-(4-iodophenyl)-5-oxovaleronitrile

  • MDL number MFCD02260577
  • Cas Number : 898767-84-5
  • Molecular Weight : 299.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6028C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6028C
Availability details for
Quantity
  • Availability
  • Add to Cart