RIE156322596 Rieke Metals, Inc.

7-(4-iodophenyl)-7-oxoheptanenitrile

  • MDL number MFCD02260579
  • Cas Number : 898767-88-9
  • Molecular Weight : 327.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6028E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6028E
Availability details for
Quantity
  • Availability
  • Add to Cart