RIE156322597 Rieke Metals, Inc.

8-(4-iodophenyl)-8-oxooctanenitrile

  • MDL number MFCD02260580
  • Cas Number : 898767-90-3
  • Molecular Weight : 341.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6028F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6028F
Availability details for
Quantity
  • Availability
  • Add to Cart