RIE156322600 Rieke Metals, Inc.

6-(3-iodophenyl)-6-oxohexanenitrile

  • MDL number MFCD02260583
  • Cas Number : 898767-95-8
  • Molecular Weight : 313.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6029D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6029D
Availability details for
Quantity
  • Availability
  • Add to Cart