RIE156322601 Rieke Metals, Inc.

7-(3-iodophenyl)-7-oxoheptanenitrile

  • MDL number MFCD02260584
  • Cas Number : 898767-97-0
  • Molecular Weight : 327.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6029E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6029E
Availability details for
Quantity
  • Availability
  • Add to Cart