RIE156322602 Rieke Metals, Inc.

8-(3-iodophenyl)-8-oxooctanenitrile

  • MDL number MFCD02260585
  • Cas Number : 898767-99-2
  • Molecular Weight : 341.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-(3-iodophenyl)-8-oxooctanenitrile
Inchi Code 1S/C14H16INO/c15-13-8-6-7-12(11-13)14(17)9-4-2-1-3-5-10-16/h6-8,11H,1-5,9H2
Inchi Key YNGAYZJNJBBQNV-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6029f

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6029f
Availability details for
Quantity
  • Availability
  • Add to Cart