RIE156322603 Rieke Metals, Inc.

4-(2-Iodophenyl)-4-oxobutyronitrile

  • MDL number MFCD02260586
  • Cas Number : 898768-01-9
  • Molecular Weight : 285.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6030B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6030B
Availability details for
Quantity
  • Availability
  • Add to Cart