RIE156322604 Rieke Metals, Inc.

6-(2-iodophenyl)-6-oxohexanenitrile

  • MDL number MFCD02260588
  • Cas Number : 898768-03-1
  • Molecular Weight : 313.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6030D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6030D
Availability details for
Quantity
  • Availability
  • Add to Cart