RIE156322607 Rieke Metals, Inc.

5-Chloro-1-(4-chlorophenyl)-1-oxopentane

  • MDL number MFCD07699516
  • Cas Number : 945-97-1
  • Molecular Weight : 231.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6031D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6031D
Availability details for
Quantity
  • Availability
  • Add to Cart