RIE156322609 Rieke Metals, Inc.

7-chloro-1-(4-chlorophenyl)-1-oxoheptane

  • MDL number MFCD07699518
  • Cas Number : 76852-66-9
  • Molecular Weight : 259.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6031F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6031F
Availability details for
Quantity
  • Availability
  • Add to Cart