RIE156322612 Rieke Metals, Inc.

6-chloro-1-(3-chlorophenyl)-1-oxohexane

  • MDL number MFCD07699522
  • Cas Number : 710339-81-4
  • Molecular Weight : 245.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6032E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6032E
Availability details for
Quantity
  • Availability
  • Add to Cart