RIE156322613 Rieke Metals, Inc.

7-chloro-1-(3-chlorophenyl)-1-oxoheptane

  • MDL number MFCD07699523
  • Cas Number : 898768-15-5
  • Molecular Weight : 259.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-chloro-1-(3-chlorophenyl)-1-heptanone
Inchi Code 1S/C13H16Cl2O/c14-9-4-2-1-3-8-13(16)11-6-5-7-12(15)10-11/h5-7,10H,1-4,8-9H2
Inchi Key GENYAAFSVKHFRS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6032f

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6032f
Availability details for
Quantity
  • Availability
  • Add to Cart