RIE156322615 Rieke Metals, Inc.

5-chloro-1-(2-chlorophenyl)-1-oxopentane

  • MDL number MFCD07699525
  • Cas Number : 487058-79-7
  • Molecular Weight : 231.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-1-(2-chlorophenyl)-1-pentanone
Inchi Code 1S/C11H12Cl2O/c12-8-4-3-7-11(14)9-5-1-2-6-10(9)13/h1-2,5-6H,3-4,7-8H2
Inchi Key DZTWKLVEJWBFKX-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6033d

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6033d
Availability details for
Quantity
  • Availability
  • Add to Cart