RIE156322617 Rieke Metals, Inc.

7-chloro-1-(2-chlorophenyl)-1-oxoheptane

  • MDL number MFCD07699527
  • Cas Number : 898768-20-2
  • Molecular Weight : 259.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-chloro-1-(2-chlorophenyl)-1-heptanone
Inchi Code 1S/C13H16Cl2O/c14-10-6-2-1-3-9-13(16)11-7-4-5-8-12(11)15/h4-5,7-8H,1-3,6,9-10H2
Inchi Key PHJYJHXZAHBZOU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6033f

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6033f
Availability details for
Quantity
  • Availability
  • Add to Cart