RIE156322618 Rieke Metals, Inc.

5-chloro-1-(4-iodophenyl)-1-oxopentane

  • MDL number MFCD07699528
  • Cas Number : 487058-91-3
  • Molecular Weight : 322.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6034D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6034D
Availability details for
Quantity
  • Availability
  • Add to Cart