RIE156322619 Rieke Metals, Inc.

6-chloro-1-(4-iodophenyl)-1-oxohexane

  • MDL number MFCD07699529
  • Cas Number : 898768-23-5
  • Molecular Weight : 336.60

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6034E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6034E
Availability details for
Quantity
  • Availability
  • Add to Cart