RIE156322626 Rieke Metals, Inc.

7-chloro-1-(2-iodophenyl)-1-oxoheptane

  • MDL number MFCD07699536
  • Cas Number : 898768-38-2
  • Molecular Weight : 350.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6036F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6036F
Availability details for
Quantity
  • Availability
  • Add to Cart