RIE156322627 Rieke Metals, Inc.

5-Chloro-1-(4-nitrophenyl)-1-oxopentane

  • MDL number MFCD08064679
  • Cas Number : 487058-76-4
  • Molecular Weight : 241.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6040D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6040D
Availability details for
Quantity
  • Availability
  • Add to Cart