RIE156322632 Rieke Metals, Inc.

7-chloro-1-(3-nitrophenyl)-1-oxoheptane

  • MDL number MFCD07699541
  • Cas Number : 898768-50-8
  • Molecular Weight : 269.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6041F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6041F
Availability details for
Quantity
  • Availability
  • Add to Cart