RIE156322635 Rieke Metals, Inc.

6-chloro-1-oxo-1-(4-trifluoromethylphenyl)hexane

  • MDL number MFCD02260643
  • Cas Number : 890043-20-6
  • Molecular Weight : 278.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6043E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6043E
Availability details for
Quantity
  • Availability
  • Add to Cart