RIE156322636 Rieke Metals, Inc.

7-chloro-1-oxo-1-(4-trifluoromethylphenyl)heptane

  • MDL number MFCD02260644
  • Cas Number : 890043-21-7
  • Molecular Weight : 292.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-chloro-1-[4-(trifluoromethyl)phenyl]-1-heptanone
Inchi Code 1S/C14H16ClF3O/c15-10-4-2-1-3-5-13(19)11-6-8-12(9-7-11)14(16,17)18/h6-9H,1-5,10H2
Inchi Key KNVAUEYSCBGRSK-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6043f

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6043f
Availability details for
Quantity
  • Availability
  • Add to Cart