RIE156322639 Rieke Metals, Inc.

6-chloro-1-oxo-1-(3-trifluoromethylphenyl)hexane

  • MDL number MFCD02260647
  • Cas Number : 898783-58-9
  • Molecular Weight : 278.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6044E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6044E
Availability details for
Quantity
  • Availability
  • Add to Cart