RIE156322640 Rieke Metals, Inc.

7-chloro-1-oxo-1-(3-trifluoromethylphenyl)heptane

  • MDL number MFCD02260648
  • Cas Number : 898783-60-3
  • Molecular Weight : 292.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6044F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6044F
Availability details for
Quantity
  • Availability
  • Add to Cart