RIE156322644 Rieke Metals, Inc.

7-chloro-1-oxo-1-(2-trifluoromethylphenyl)heptane

  • MDL number MFCD02260652
  • Cas Number : 898783-68-1
  • Molecular Weight : 292.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6045F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6045F
Availability details for
Quantity
  • Availability
  • Add to Cart