RIE156322647 Rieke Metals, Inc.

6-oxo-6-(4-trifluoromethylphenyl)hexanenitrile

  • MDL number MFCD02260655
  • Cas Number : 61718-88-5
  • Molecular Weight : 255.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-oxo-6-[4-(trifluoromethyl)phenyl]hexanenitrile
Inchi Code 1S/C13H12F3NO/c14-13(15,16)11-7-5-10(6-8-11)12(18)4-2-1-3-9-17/h5-8H,1-4H2
Inchi Key UEGQWUAFHTVABM-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6046d

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6046d
Availability details for
Quantity
  • Availability
  • Add to Cart