RIE156322648 Rieke Metals, Inc.

7-oxo-7-(4-trifluoromethylphenyl)heptanenitrile

  • MDL number MFCD02260656
  • Cas Number : 898783-72-7
  • Molecular Weight : 269.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-oxo-7-[4-(trifluoromethyl)phenyl]heptanenitrile
Inchi Code 1S/C14H14F3NO/c15-14(16,17)12-8-6-11(7-9-12)13(19)5-3-1-2-4-10-18/h6-9H,1-5H2
Inchi Key AMGBXJDSVRZTET-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6046e

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6046e
Availability details for
Quantity
  • Availability
  • Add to Cart