RIE156322649 Rieke Metals, Inc.

8-oxo-8-(4-trifluoromethylphenyl)octanenitrile

  • MDL number MFCD02260657
  • Cas Number : 898783-74-9
  • Molecular Weight : 283.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-oxo-8-[4-(trifluoromethyl)phenyl]octanenitrile
Inchi Code 1S/C15H16F3NO/c16-15(17,18)13-9-7-12(8-10-13)14(20)6-4-2-1-3-5-11-19/h7-10H,1-6H2
Inchi Key DAFXTBXSBIZTRX-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6046f

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6046f
Availability details for
Quantity
  • Availability
  • Add to Cart