RIE156322650 Rieke Metals, Inc.

5-(4-Chlorophenyl)-5-oxovaleronitrile

  • MDL number MFCD02260658
  • Cas Number : 54959-85-2
  • Molecular Weight : 207.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6047C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6047C
Availability details for
Quantity
  • Availability
  • Add to Cart