RIE156322655 Rieke Metals, Inc.

2'-chloro-2-phenylacetophenone

  • MDL number MFCD02260664
  • Cas Number : 72867-72-2
  • Molecular Weight : 230.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6050-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6050-1
Availability details for
Quantity
  • Availability
  • Add to Cart