RIE156322657 Rieke Metals, Inc.

4'-Iodo-2-phenylacetophenone

  • MDL number MFCD01027442
  • Cas Number : 55794-28-0
  • Molecular Weight : 322.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6052-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6052-3
Availability details for
Quantity
  • Availability
  • Add to Cart