RIE156322659 Rieke Metals, Inc.

3'-Cyano-2-phenylacetophenone

  • MDL number MFCD02260674
  • Cas Number : 758710-68-8
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6055-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6055-2
Availability details for
Quantity
  • Availability
  • Add to Cart