RIE156322660 Rieke Metals, Inc.

2-phenyl-2'-trifluoromethylacetophenone

  • MDL number MFCD02260676
  • Cas Number : 898783-80-7
  • Molecular Weight : 264.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6056-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6056-1
Availability details for
Quantity
  • Availability
  • Add to Cart