RIE156322662 Rieke Metals, Inc.

2-(3-Methylphenyl)acetophenone

  • MDL number MFCD02260689
  • Cas Number : 34403-03-7
  • Molecular Weight : 210.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6061-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6061-2
Availability details for
Quantity
  • Availability
  • Add to Cart