RIE156322666 Rieke Metals, Inc.

2-(3-bromophenyl)-2'-iodoacetophenone

  • MDL number MFCD02260696
  • Cas Number : 898783-88-5
  • Molecular Weight : 401.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6065-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6065-1
Availability details for
Quantity
  • Availability
  • Add to Cart