RIE156322675 Rieke Metals, Inc.

2-(3-chlorophenyl)-2'-iodoacetophenone

  • MDL number MFCD02260705
  • Cas Number : 898784-03-7
  • Molecular Weight : 356.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6068-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6068-1
Availability details for
Quantity
  • Availability
  • Add to Cart