RIE156322681 Rieke Metals, Inc.

2-(2-bromophenyl)-2'-cyanoacetophenone

  • MDL number MFCD02260720
  • Cas Number : 898784-09-3
  • Molecular Weight : 300.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-[(2-bromophenyl)acetyl]benzonitrile
Inchi Code 1S/C15H10BrNO/c16-14-8-4-2-5-11(14)9-15(18)13-7-3-1-6-12(13)10-17/h1-8H,9H2
Inchi Key RQVORDZXQUNEHT-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6073-1

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6073-1
Availability details for
Quantity
  • Availability
  • Add to Cart