RIE156322683 Rieke Metals, Inc.

2-(2-bromophenyl)-4'-cyanoacetophenone

  • MDL number MFCD02260722
  • Cas Number : 898784-11-7
  • Molecular Weight : 300.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-[(2-bromophenyl)acetyl]benzonitrile
Inchi Code 1S/C15H10BrNO/c16-14-4-2-1-3-13(14)9-15(18)12-7-5-11(10-17)6-8-12/h1-8H,9H2
Inchi Key XFWFAQYYNUKYEQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6073-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6073-3
Availability details for
Quantity
  • Availability
  • Add to Cart