RIE156322684 Rieke Metals, Inc.

2-(3-bromophenyl)-3'-cyanoacetophenone

  • MDL number MFCD02260724
  • Cas Number : 898784-12-8
  • Molecular Weight : 300.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-[(3-bromophenyl)acetyl]benzonitrile
Inchi Code 1S/C15H10BrNO/c16-14-6-2-3-11(8-14)9-15(18)13-5-1-4-12(7-13)10-17/h1-8H,9H2
Inchi Key NTKNEVRHLMMHCA-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/6074-2

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6074-2
Availability details for
Quantity
  • Availability
  • Add to Cart