RIE156322685 Rieke Metals, Inc.

2-(3-bromophenyl)-4'-cyanoacetophenone

  • MDL number MFCD02260725
  • Cas Number : 898784-13-9
  • Molecular Weight : 300.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/6074-3

Safety Information

MSDS http://riekemetals.com/products-services/catnum/6074-3
Availability details for
Quantity
  • Availability
  • Add to Cart